Assay Details
| Gene Symbol: WT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000378.4 | NP_000369.3 | 4 - 5 | 1146 | 65 | |
| NM_001198552.1 | NP_001185481.1 | 4 - 5 | 598 | 65 | ||
| GenBank mRNA | AK093168.1 | - | 4 - 5 | 594 | 65 | |
| M30393.1 | - | 4 - 5 | 500 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

WT1
Genomic Map