Assay Details
| Gene Symbol: ANKLE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015114.2 | NP_055929.1 | 3 - 4 | 944 | 76 | |
| XM_005266159.3 | XP_005266216.1 | 3 - 4 | 865 | 76 | ||
| XM_005266160.1 | XP_005266217.1 | 3 - 4 | 801 | 76 | ||
| XM_006719735.1 | XP_006719798.1 | 3 - 4 | 855 | 76 | ||
| XM_011534787.2 | XP_011533089.1 | 3 - 4 | 855 | 76 | ||
| XM_011534788.1 | XP_011533090.1 | 3 - 4 | 855 | 76 | ||
| XM_017019076.1 | XP_016874565.1 | 3 - 4 | 801 | 76 | ||
| GenBank mRNA | AB014592.1 | - | 2 - 3 | 391 | 76 | |
| BC001530.2 | - | 2 - 3 | 233 | 76 | ||
| BC025347.1 | - | 2 - 3 | 233 | 76 | ||
| BC043157.1 | - | 3 - 4 | 890 | 5286234 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002155.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017019076.1 transcripts.
Set Membership: |
|

ANKLE2
Genomic Map