Assay Details
| Gene Symbol: SLC6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003042.3 | NP_003033.3 | 3 - 4 | 645 | 80 | |
| XM_005265410.4 | XP_005265467.1 | 3 - 4 | 2002 | 80 | ||
| XM_005265411.4 | XP_005265468.1 | 3 - 4 | 1941 | 80 | ||
| XM_006713306.3 | XP_006713369.1 | 3 - 4 | 600 | 80 | ||
| XM_011534025.2 | XP_011532327.1 | 3 - 4 | 2692 | 80 | ||
| XM_011534027.2 | XP_011532329.1 | 3 - 4 | 2631 | 80 | ||
| XM_017007071.1 | XP_016862560.1 | 4 - 5 | 2424 | 80 | ||
| XM_017007072.1 | XP_016862561.1 | 4 - 5 | 1734 | 80 | ||
| GenBank mRNA | BC033904.1 | - | 3 - 4 | 613 | 5264672 | 80 |
| BC057291.1 | - | 3 - 4 | 625 | 80 | ||
| X54673.1 | - | 2 - 3 | 468 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000063.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
neurotransmitter transport
chemical synaptic transmission
learning
response to toxic substance
response to sucrose
response to lead ion
positive regulation of gamma-aminobutyric acid secretion
response to purine-containing compound
gamma-aminobutyric acid transport
negative regulation of synaptic transmission, GABAergic
response to estradiol
response to cocaine
protein homooligomerization
response to calcium ion
gamma-aminobutyric acid import
transmembrane transport
mitophagy in response to mitochondrial depolarization
neurotransmitter:sodium symporter activity
gamma-aminobutyric acid:sodium symporter activity
metal ion binding
transport
neurotransmitter transport
chemical synaptic transmission
learning
response to toxic substance
response to sucrose
response to lead ion
positive regulation of gamma-aminobutyric acid secretion
response to purine-containing compound
gamma-aminobutyric acid transport
negative regulation of synaptic transmission, GABAergic
response to estradiol
response to cocaine
protein homooligomerization
response to calcium ion
gamma-aminobutyric acid import
transmembrane transport
mitophagy in response to mitochondrial depolarization
neurotransmitter:sodium symporter activity
gamma-aminobutyric acid:sodium symporter activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03001375.hg.1 | TC03000063.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001375.hg.1 | ENST00000287766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001375.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001376.hg.1 | TC03000063.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001376.hg.1 | ENST00000287766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001376.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001376.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001377.hg.1 | TC03000063.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001377.hg.1 | ENST00000287766.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03001377.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000460480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000460480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000460480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000460480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000460480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000495636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000495636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000495636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000495636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | ENST00000495636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300145797.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300145797.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300145797.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300145797.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300145797.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300145797.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300145797.hg.1 | TC0300006599.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300145798.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300145798.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300145798.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300145798.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300145798.hg.1 | TC0300006599.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000425938.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000460480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000460480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000460480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000460480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000460480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000462473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000495636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000495636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000495636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000495636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | ENST00000495636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300006599.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300145796.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300145796.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300145796.hg.1 | ENST00000287766.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300145796.hg.1 | NM_003042.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300145796.hg.1 | TC0300006599.hg.1 | Clariom D TC |

SLC6A1
Genomic Map