Assay Details
| Gene Symbol: ATP9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006045.2 | NP_006036.1 | 25 - 26 | 2788 | 104 | |
| XM_011528481.2 | XP_011526783.1 | 25 - 26 | 2816 | 104 | ||
| GenBank mRNA | AB014511.1 | - | 20 - 21 | 2342 | 104 | |
| AK172802.1 | - | 23 - 24 | 2477 | 104 | ||
| AK299064.1 | - | 25 - 26 | 2717 | 104 | ||
| BC110591.1 | - | 21 - 22 | 2280 | 104 | ||
| BC110592.1 | - | 21 - 22 | 2280 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000944.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20013019.hg.1 | ENST00000311637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013019.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013019.hg.1 | ENST00000402822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013019.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013019.hg.1 | TC20000944.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013018.hg.1 | ENST00000311637.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013018.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013018.hg.1 | ENST00000402822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013018.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20013018.hg.1 | TC20000944.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | ENST00000477492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009461.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166155.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166155.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166155.hg.1 | NM_006045.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000166155.hg.1 | TC2000009461.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000166155.hg.1 | ENST00000311637.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166155.hg.1 | ENST00000338821.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000166155.hg.1 | NM_006045.2 | Refseq |

ATP9A
Genomic Map