Assay Details
| Gene Symbol: USP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199443.2 | NP_955475.1 | 6 - 7 | 779 | 62 | |
| GenBank mRNA | AF017306.1 | - | 6 - 7 | 700 | 62 | |
| AK222725.1 | - | 6 - 7 | 709 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003383.hg.1
Set Membership: |
|

USP4
Genomic Map