Assay Details
| Gene Symbol: PARP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022750.3 | NP_073587.1 | 8 - 9 | 2302 | 80 | |
| NR_130117.1 | 7 - 8 | 2160 | 80 | |||
| XM_011516491.2 | XP_011514793.1 | 8 - 9 | 2297 | 80 | ||
| GenBank mRNA | AK026346.1 | - | 2 - 3 | 142 | 80 | |
| AL136766.1 | - | 8 - 9 | 1924 | 80 | ||
| AL137255.1 | - | 8 - 9 | 1121 | 80 | ||
| BC081541.1 | - | 8 - 9 | 2302 | 5296560 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001916.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000263549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000466597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000466597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000466597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000466597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000466597.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000467638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000467638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000467638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000470515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000470515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000470515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000470515.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000473341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000484111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000484111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000484111.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000488726.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000489809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000491598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000491598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000491598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000491598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000491598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000491598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000491598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000493262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000493262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000493262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000493262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000493262.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000496624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000496624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000496624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000496624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | ENST00000496624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NM_022750.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012797.hg.1 | NR_130117.1 | Refseq |

PARP12
Genomic Map