Assay Details
| Gene Symbol: SLC9B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300754.1 | NP_001287683.1 | 3 - 4 | 537 | 98 | |
| NM_001300756.1 | NP_001287685.1 | 3 - 4 | 903 | 98 | ||
| XM_006714086.3 | XP_006714149.1 | 3 - 4 | 902 | 98 | ||
| GenBank mRNA | AK297010.1 | - | 3 - 4 | 448 | 98 | |
| AK299888.1 | - | 3 - 4 | 462 | 98 | ||
More Information
Set Membership: |
|

SLC9B2
Genomic Map