Assay Details
| Gene Symbol: ZNFX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021035.2 | NP_066363.1 | 5 - 6 | 2404 | 66 | |
| XM_006723836.1 | XP_006723899.1 | 5 - 6 | 2234 | 66 | ||
| GenBank mRNA | AB037825.2 | - | 5 - 6 | 2222 | 66 | |
| AK002139.1 | - | 5 - 6 | 2317 | 66 | ||
| AK022641.1 | - | 3 - 4 | 1569 | 66 | ||
| AK023836.1 | - | 1 - 2 | 156 | 66 | ||
| AK307392.1 | - | 5 - 6 | 2608 | 66 | ||
| BC014041.2 | - | 4 - 5 | 2220 | 66 | ||
| DA809204.1 | - | 3 - 4 | 497 | 66 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000924.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012734.hg.1 | ENST00000371752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012734.hg.1 | ENST00000371754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012734.hg.1 | ENST00000396105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012734.hg.1 | ENST00000396106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012734.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012734.hg.1 | TC20000924.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171800.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171800.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171800.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171800.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171800.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171800.hg.1 | TC2000010022.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171800.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171800.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171800.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171800.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171800.hg.1 | NM_021035.2 | Refseq |

ZNFX1
Genomic Map