Assay Details
| Gene Symbol: ZNFX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021035.2 | NP_066363.1 | 12 - 13 | 3460 | 106 | |
| XM_006723836.1 | XP_006723899.1 | 12 - 13 | 3290 | 106 | ||
| XM_011528924.1 | XP_011527226.1 | 11 - 12 | 3335 | 106 | ||
| XM_017027971.1 | XP_016883460.1 | 10 - 11 | 2386 | 106 | ||
| GenBank mRNA | AB037825.2 | - | 12 - 13 | 3278 | 106 | |
| AK023836.1 | - | 8 - 9 | 1212 | 106 | ||
| BC041630.1 | - | 5 - 6 | 744 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000924.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012723.hg.1 | ENST00000371752.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012723.hg.1 | ENST00000371754.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012723.hg.1 | ENST00000396105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012723.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20012723.hg.1 | TC20000924.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000171791.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171791.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171791.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171791.hg.1 | TC2000010022.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171791.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171791.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171791.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371744.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000371752.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000396105.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | ENST00000455070.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010022.hg.1 | NM_021035.2 | Refseq |

ZNFX1
Genomic Map