Assay Details
| Gene Symbol: HAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079870.1 | NP_001073339.1 | 8 - 9 | 1323 | 120 | |
| NM_001079871.1 | NP_001073340.1 | 8 - 9 | 1299 | 120 | ||
| GenBank mRNA | AB209105.1 | - | 8 - 9 | 1284 | 120 | |
| AK022007.1 | - | 8 - 9 | 1320 | 120 | ||
More Information
Set Membership: |
|

HAP1
Genomic Map