Assay Details
| Gene Symbol: TAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013998.2 | NP_054704.1 | 3 - 4 | 464 | 83 | |
| GenBank mRNA | AF050658.1 | - | 2 - 3 | 113 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013998.2 | NP_054704.1 | 3 - 4 | 464 | 83 | |
| GenBank mRNA | AF050658.1 | - | 2 - 3 | 113 | 83 | |
Set Membership: |
|