Assay Details
| Gene Symbol: TAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013998.2 | NP_054704.1 | 4 - 5 | 490 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000579.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of acute inflammatory response
inflammatory response
positive regulation of cytosolic calcium ion concentration
tachykinin receptor signaling pathway
neuropeptide signaling pathway
cell-cell signaling
chemical synaptic transmission
insemination
long-term memory
regulation of blood pressure
associative learning
detection of abiotic stimulus
response to hormone
negative regulation of heart rate
positive regulation of epithelial cell migration
sensory perception of pain
positive regulation of synaptic transmission, cholinergic
positive regulation of synaptic transmission, GABAergic
response to lipopolysaccharide
positive regulation of renal sodium excretion
response to morphine
positive regulation of action potential
positive regulation of ossification
positive regulation of saliva secretion
response to pain
positive regulation of lymphocyte proliferation
positive regulation of stress fiber assembly
cellular response to nerve growth factor stimulus
positive regulation of corticosterone secretion
receptor binding
substance P receptor binding
positive regulation of acute inflammatory response
inflammatory response
positive regulation of cytosolic calcium ion concentration
tachykinin receptor signaling pathway
neuropeptide signaling pathway
cell-cell signaling
chemical synaptic transmission
insemination
long-term memory
regulation of blood pressure
associative learning
detection of abiotic stimulus
response to hormone
negative regulation of heart rate
positive regulation of epithelial cell migration
sensory perception of pain
positive regulation of synaptic transmission, cholinergic
positive regulation of synaptic transmission, GABAergic
response to lipopolysaccharide
positive regulation of renal sodium excretion
response to morphine
positive regulation of action potential
positive regulation of ossification
positive regulation of saliva secretion
response to pain
positive regulation of lymphocyte proliferation
positive regulation of stress fiber assembly
cellular response to nerve growth factor stimulus
positive regulation of corticosterone secretion
receptor binding
substance P receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000491437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000491437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000491437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000491437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000495916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000495916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | ENST00000495916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008478.hg.1 | TC07000579.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008478.hg.1 | ENST00000319273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008478.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008478.hg.1 | ENST00000350485.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008478.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008478.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008478.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008478.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | ENST00000491437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | ENST00000491437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158849.hg.1 | TC0700008435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158850.hg.1 | TC0700008435.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000319273.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000346867.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000350485.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000491437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000491437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000491437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000491437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000495916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000495916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | ENST00000495916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_003182.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013996.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013997.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013998.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008435.hg.1 | NM_013998.2 | Refseq |

TAC1
Genomic Map