Assay Details
| Gene Symbol: PCP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BQ287895.1 | - | 1 - 2 | 418 | 97 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000328619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000328619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000328619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000328619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000328619.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000462224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000462224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000462224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000462224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000462224.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000467565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000467565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000467565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000467565.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000468717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000468717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000468717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000468717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | ENST00000468717.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | NM_006198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | NM_006198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | NM_006198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007176.hg.1 | NM_006198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000328619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000328619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000328619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000328619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000328619.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000462224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000462224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000462224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000462224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000462224.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000467565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000467565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000467565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000467565.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000468717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000468717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000468717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000468717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | ENST00000468717.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | NM_006198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | NM_006198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | NM_006198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007176.hg.1 | NM_006198.2 | Refseq |

PCP4
Genomic Map