| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016135.hg.1 |
ENST00000268717.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016135.hg.1 |
ENST00000486810.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016135.hg.1 |
ENST00000539941.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016135.hg.1 |
NM_001199125.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016135.hg.1 |
NM_003653.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17016135.hg.1 |
TC17001186.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000268717.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000417352.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000439936.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000462236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000477299.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000486810.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000492672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000495640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000539941.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000577246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000578317.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000583160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
NM_001199125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
NM_003653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000268717.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000417352.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000439936.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000462236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000477299.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000486810.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000492672.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000495640.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000539941.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000577246.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000578317.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
ENST00000583160.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
NM_001199125.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
NM_003653.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700177643.hg.1 |
TC1700009928.hg.1 |
Clariom D TC |