Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014057.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014057.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014057.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014057.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014057.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014057.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014057.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014057.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014057.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014057.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014057.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014057.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014057.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014057.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014057.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014057.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205131.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205131.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205131.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205131.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205131.hg.1 |
TC0100014057.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100205132.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205132.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205132.hg.1 |
ENST00000416581.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100205132.hg.1 |
NR_121680.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100205132.hg.1 |
TC0100014057.hg.1 |
Clariom D TC |