Assay Details
| Gene Symbol: SRGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005269042.3 | XP_005269099.1 | 11 - 12 | 1881 | 71 | |
| GenBank mRNA | AB037725.1 | - | 10 - 11 | 2783 | 71 | |
More Information
Set Membership: |
|

SRGAP1
Genomic Map