Assay Details
| Gene Symbol: ADAM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288973.1 | NP_001275902.1 | 17 - 18 | 2408 | 65 | |
| NM_001288974.1 | NP_001275903.1 | 17 - 18 | 2408 | 65 | ||
| NM_001288975.1 | NP_001275904.1 | 17 - 18 | 2408 | 65 | ||
| NM_003474.5 | NP_003465.3 | 17 - 18 | 2417 | 65 | ||
| NM_021641.4 | NP_067673.2 | 17 - 18 | 2417 | 65 | ||
| XM_017016705.1 | XP_016872194.1 | 16 - 17 | 2320 | 65 | ||
| XM_017016706.1 | XP_016872195.1 | 7 - 8 | 1459 | 65 | ||
| GenBank mRNA | AF023476.2 | - | 17 - 18 | 2310 | 65 | |
| AF023477.2 | - | 17 - 18 | 2310 | 65 | ||
| AK127856.1 | - | 17 - 18 | 2313 | 65 | ||
| AK291629.1 | - | 17 - 18 | 2227 | 65 | ||
| AY358878.1 | - | 17 - 18 | 2305 | 65 | ||
| BC060804.1 | - | 17 - 18 | 2347 | 30348514 | 65 | |
| KF444157.1 | - | 17 - 18 | 2408 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001744.hg.1
Set Membership: |
|

ADAM12
Genomic Map