Assay Details
| Gene Symbol: PHGDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006623.3 | NP_006614.2 | 3 - 4 | 585 | 80 | |
| GenBank mRNA | AF171237.1 | - | 3 - 4 | 431 | 80 | |
| AK093306.1 | - | 2 - 3 | 1763 | 80 | ||
| AK315360.1 | - | 3 - 4 | 506 | 80 | ||
| BC000303.2 | - | 3 - 4 | 425 | 2820369 | 80 | |
| BC001349.2 | - | 3 - 4 | 420 | 2961389 | 80 | |
| BC011262.1 | - | 3 - 4 | 494 | 4156703 | 80 | |
| BC023235.1 | - | 3 - 4 | 398 | 80 | ||
| CR456795.1 | - | 3 - 4 | 358 | 80 | ||
| FJ224347.1 | - | 1050 | 80 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001043.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glutamine metabolic process
glycine metabolic process
L-serine biosynthetic process
threonine metabolic process
brain development
serine family amino acid biosynthetic process
gamma-aminobutyric acid metabolic process
regulation of gene expression
taurine metabolic process
spinal cord development
glial cell development
neural tube development
neuron projection development
oxidation-reduction process
G1 to G0 transition
phosphoglycerate dehydrogenase activity
electron carrier activity
NAD binding
glutamine metabolic process
glycine metabolic process
L-serine biosynthetic process
threonine metabolic process
brain development
serine family amino acid biosynthetic process
gamma-aminobutyric acid metabolic process
regulation of gene expression
taurine metabolic process
spinal cord development
glial cell development
neural tube development
neuron projection development
oxidation-reduction process
G1 to G0 transition
phosphoglycerate dehydrogenase activity
electron carrier activity
NAD binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01016518.hg.1 | TC01001043.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016518.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016518.hg.1 | ENST00000369409.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016518.hg.1 | ENST00000535091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016518.hg.1 | ENST00000537497.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016518.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000469443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000469443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000469443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000469443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000496756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000496756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000496756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000496756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | ENST00000496756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000469443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000469443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000469443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000469443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000482968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000496756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000496756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000496756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000496756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | ENST00000496756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009621.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169256.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169256.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169256.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169256.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169256.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169256.hg.1 | ENST00000369407.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169256.hg.1 | ENST00000369409.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169256.hg.1 | ENST00000462324.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169256.hg.1 | ENST00000493622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100169256.hg.1 | NM_006623.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100169256.hg.1 | TC0100009621.hg.1 | Clariom D TC |

PHGDH
Genomic Map