Assay Details
| Gene Symbol: CABLES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100619.2 | NP_001094089.1 | 7 - 8 | 1442 | 66 | |
| NM_001256438.1 | NP_001243367.1 | 7 - 8 | 704 | 66 | ||
| NM_138375.2 | NP_612384.1 | 7 - 8 | 725 | 66 | ||
| GenBank mRNA | AF348525.1 | - | 7 - 8 | 842 | 66 | |
| AK296407.1 | - | 7 - 8 | 704 | 66 | ||
| AK304880.1 | - | 4 - 5 | 389 | 66 | ||
| BC004124.2 | - | 2 - 3 | 221 | 66 | ||
| BC037218.1 | - | 7 - 8 | 725 | 5196987 | 66 | |
| EF028204.1 | - | 7 - 8 | 1442 | 66 | ||
More Information
Set Membership: |
|

CABLES1
Genomic Map