Assay Details
| Gene Symbol: MS4A4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243266.1 | NP_001230195.1 | 3 - 4 | 453 | 133 | |
| NM_024021.3 | NP_076926.2 | 4 - 5 | 599 | 133 | ||
| NM_148975.2 | NP_683876.1 | 3 - 4 | 453 | 133 | ||
| XM_017017909.1 | XP_016873398.1 | 3 - 4 | 466 | 133 | ||
| GenBank mRNA | AB013102.1 | - | 3 - 4 | 294 | 133 | |
| AB022821.1 | - | 3 - 4 | 318 | 133 | ||
| AF068288.1 | - | 3 - 4 | 387 | 133 | ||
| AF237912.1 | - | 3 - 4 | 404 | 133 | ||
| AF350500.1 | - | 2 - 3 | 357 | 133 | ||
| AF354928.1 | - | 2 - 3 | 277 | 133 | ||
| AK292089.1 | - | 3 - 4 | 408 | 133 | ||
| AK314033.1 | - | 3 - 4 | 296 | 133 | ||
| BC020648.1 | - | 3 - 4 | 328 | 4720088 | 133 | |
| CB046393.1 | - | 3 - 4 | 322 | 133 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000506.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11006008.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006008.hg.1 | ENST00000355131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006008.hg.1 | ENST00000395016.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006008.hg.1 | ENST00000532114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006008.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006008.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006008.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006008.hg.1 | TC11000506.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000529991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000529991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000529991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000529991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100154712.hg.1 | TC1100007767.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000337908.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000343968.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000527056.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000529950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000529991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000529991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000529991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000529991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | ENST00000532114.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_001243266.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_024021.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007767.hg.1 | NM_148975.2 | Refseq |

MS4A4A
Genomic Map