Assay Details
| Gene Symbol: THOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081550.1 | NP_001075019.1 | 37 - 38 | 4792 | 86 | |
| XM_017029662.1 | XP_016885151.1 | 39 - 40 | 5104 | 86 | ||
| XM_017029666.1 | XP_016885155.1 | 39 - 40 | 5092 | 86 | ||
| XM_017029667.1 | XP_016885156.1 | 39 - 40 | 5089 | 86 | ||
| XM_017029669.1 | XP_016885158.1 | 38 - 39 | 5038 | 86 | ||
| XM_017029670.1 | XP_016885159.1 | 38 - 39 | 5026 | 86 | ||
| XM_017029671.1 | XP_016885160.1 | 38 - 39 | 5023 | 86 | ||
| XM_017029672.1 | XP_016885161.1 | 39 - 40 | 5008 | 86 | ||
| XM_017029673.1 | XP_016885162.1 | 38 - 39 | 4966 | 86 | ||
| XM_017029674.1 | XP_016885163.1 | 38 - 39 | 4954 | 86 | ||
| XM_017029675.1 | XP_016885164.1 | 38 - 39 | 4942 | 86 | ||
| XM_017029677.1 | XP_016885166.1 | 37 - 38 | 4900 | 86 | ||
| XM_017029678.1 | XP_016885167.1 | 37 - 38 | 4888 | 86 | ||
| XM_017029679.1 | XP_016885168.1 | 37 - 38 | 4804 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

THOC2
Genomic Map