Assay Details
| Gene Symbol: ZNF670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204220.1 | NP_001191149.1 | 1 - 2 | 221 | 58 | |
| NM_033213.4 | NP_149990.1 | 1 - 2 | 221 | 58 | ||
| GenBank mRNA | AK074774.1 | - | 1 - 2 | 168 | 58 | |
| BG532425.1 | - | 1 - 2 | 175 | 58 | ||
| DA757117.1 | - | 1 - 2 | 221 | 58 | ||
| DB291578.1 | - | 1 - 2 | 169 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK074774.1, NM_001204220.1, NM_033213.4 transcripts.
Set Membership: |
|

ZNF670
Genomic Map