Assay Details
| Gene Symbol: PITPNM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165966.1 | NP_001159438.1 | 2 - 3 | 265 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PITPNM3| Gene Symbol: PITPNM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165966.1 | NP_001159438.1 | 2 - 3 | 265 | 77 | |
Set Membership: |
|