Assay Details
| Gene Symbol: LGALSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014181.2 | NP_054900.2 | 3 - 4 | 547 | 56 | |
| GenBank mRNA | AF161508.1 | - | 3 - 4 | 218 | 56 | |
| AK314656.1 | - | 3 - 4 | 248 | 56 | ||
| BC036082.1 | - | 3 - 4 | 219 | 5301908 | 56 | |
| BC062691.1 | - | 3 - 4 | 264 | 6451913 | 56 | |
| DR001728.1 | - | 3 - 4 | 555 | 56 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000366.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005615.hg.1 | TC02000366.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005615.hg.1 | ENST00000238875.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005615.hg.1 | ENST00000464281.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02005615.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153861.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153861.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153861.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153861.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153861.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153861.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153861.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153861.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153861.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153861.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153861.hg.1 | TC0200007803.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000238875.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000409537.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000420552.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000462737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | ENST00000464281.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007803.hg.1 | NM_014181.2 | Refseq |

LGALSL
Genomic Map