Assay Details
| Gene Symbol: BIRC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016252.3 | NP_057336.3 | 57 - 58 | 11753 | 158 | |
| XM_005264449.4 | XP_005264506.2 | 57 - 58 | 11955 | 158 | ||
| XM_005264450.4 | XP_005264507.2 | 57 - 58 | 11943 | 158 | ||
| XM_005264451.4 | XP_005264508.2 | 57 - 58 | 11928 | 158 | ||
| XM_005264452.4 | XP_005264509.2 | 57 - 58 | 11658 | 158 | ||
| XM_005264453.4 | XP_005264510.2 | 57 - 58 | 11631 | 158 | ||
| XM_005264454.4 | XP_005264511.2 | 57 - 58 | 11955 | 158 | ||
| XM_005264455.4 | XP_005264512.2 | 56 - 57 | 11823 | 158 | ||
| XM_006712054.3 | XP_006712117.1 | 57 - 58 | 11937 | 158 | ||
| XM_006712055.3 | XP_006712118.1 | 57 - 58 | 11895 | 158 | ||
| XM_006712056.3 | XP_006712119.1 | 57 - 58 | 11856 | 158 | ||
| XM_011533003.2 | XP_011531305.1 | 57 - 58 | 11400 | 158 | ||
| XM_017004556.1 | XP_016860045.1 | 57 - 58 | 11877 | 158 | ||
| XM_017004557.1 | XP_016860046.1 | 57 - 58 | 11844 | 158 | ||
| XM_017004558.1 | XP_016860047.1 | 57 - 58 | 11814 | 158 | ||
| XM_017004559.1 | XP_016860048.1 | 57 - 58 | 11955 | 158 | ||
| GenBank mRNA | AB033115.2 | - | 28 - 29 | 5433 | 158 | |
| AF265555.1 | - | 57 - 58 | 11535 | 158 | ||
| AK023848.1 | - | 12 - 13 | 2705 | 158 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000213.hg.1
Set Membership: |
|

BIRC6
Genomic Map