Assay Details
| Gene Symbol: NR1H3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005252709.1 | XP_005252766.1 | 3 - 4 | 244 | 62 | |
| XM_005252710.1 | XP_005252767.1 | 2 - 3 | 142 | 62 | ||
| XM_006718115.1 | XP_006718178.1 | 2 - 3 | 250 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005252709.1, XM_005252710.1, XM_006718115.1 transcripts.
Set Membership: |
|

NR1H3
Genomic Map