Assay Details
| Gene Symbol: SLC20A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257180.1 | NP_001244109.1 | 2 - 3 | 989 | 87 | |
| NM_001257181.1 | NP_001244110.1 | 2 - 3 | 620 | 87 | ||
| NM_006749.4 | NP_006740.1 | 2 - 3 | 952 | 87 | ||
| XM_005273613.3 | XP_005273670.1 | 2 - 3 | 727 | 87 | ||
| XM_005273615.3 | XP_005273672.1 | 2 - 3 | 727 | 87 | ||
| XM_017013748.1 | XP_016869237.1 | 2 - 3 | 843 | 87 | ||
| GenBank mRNA | AK291202.1 | - | 2 - 3 | 989 | 87 | |
| AK314774.1 | - | 2 - 3 | 621 | 87 | ||
| BC028600.2 | - | 2 - 3 | 664 | 4820601 | 87 | |
| L20852.1 | - | 1 - 2 | 538 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001179.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
phosphate ion transmembrane transport
sodium ion transmembrane transport
sodium-dependent phosphate transport
viral entry into host cell
virus receptor activity
receptor activity
inorganic phosphate transmembrane transporter activity
sodium:phosphate symporter activity
sodium:inorganic phosphate symporter activity
sodium-dependent phosphate transmembrane transporter activity
transport
ion transport
phosphate ion transmembrane transport
sodium ion transmembrane transport
sodium-dependent phosphate transport
viral entry into host cell
virus receptor activity
receptor activity
inorganic phosphate transmembrane transporter activity
sodium:phosphate symporter activity
sodium:inorganic phosphate symporter activity
sodium-dependent phosphate transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000342228.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000517366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000518717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000519463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000519463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000519463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520179.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000520262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000521455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000521455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000521455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000521455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522401.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000522707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000523340.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | ENST00000524237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257180.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_001257181.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010285.hg.1 | NM_006749.4 | Refseq |

SLC20A2
Genomic Map