Assay Details
| Gene Symbol: PAK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011526316.1 | XP_011524618.1 | 2 - 3 | 212 | 78 | |
| XM_011526320.1 | XP_011524622.1 | 2 - 3 | 199 | 78 | ||
| GenBank mRNA | AX775871.1 | - | 2 - 3 | 200 | 78 | |
| BC034511.1 | - | 2 - 3 | 200 | 3882622 | 78 | |
| BG773480.1 | - | 2 - 3 | 244 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AX775871.1, BC034511.1, XM_011526316.1, XM_011526320.1 transcripts.
Set Membership: |
|

PAK4
Genomic Map