Assay Details
| Gene Symbol: RCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005259089.3 | XP_005259146.1 | 2 - 3 | 637 | 75 | |
| XM_011527143.1 | XP_011525445.1 | 2 - 3 | 637 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005259089.3, XM_011527143.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000593483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000593483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000593483.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000593644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000593644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000593644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000270645.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000593483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000593483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000593483.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000593644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000593644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000593644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000597801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | ENST00000598833.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008544.hg.1 | NM_020650.2 | Refseq |

RCN3
Genomic Map