Assay Details
| Gene Symbol: CHERP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006387.5 | NP_006378.3 | 2 - 3 | 269 | 96 | |
| GenBank mRNA | AF536542.1 | - | 2 - 3 | 236 | 96 | |
| AK296883.1 | - | 2 - 3 | 248 | 96 | ||
| BC001552.2 | - | 2 - 3 | 227 | 96 | ||
| BC021294.2 | - | 2 - 3 | 254 | 4053575 | 96 | |
| CN336425.1 | - | 2 - 3 | 268 | 96 | ||
| U94836.2 | - | 1 - 2 | 171 | 96 | ||
| Y08265.1 | - | 2 - 3 | 226 | 96 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19002687.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK296883.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
cellular calcium ion homeostasis
nervous system development
negative regulation of cell proliferation
release of sequestered calcium ion into cytosol
positive regulation of NFAT protein import into nucleus
protein binding
ion channel binding
poly(A) RNA binding
RNA processing
cellular calcium ion homeostasis
nervous system development
negative regulation of cell proliferation
release of sequestered calcium ion into cytosol
positive regulation of NFAT protein import into nucleus
protein binding
ion channel binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19017842.hg.1 | TC19002687.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017842.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017842.hg.1 | ENST00000546361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017842.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000551747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000551747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000551747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000597261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000597261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000597261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000597261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000597261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000600432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000600432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000600432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | ENST00000600432.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900204978.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204978.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204978.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204978.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900204978.hg.1 | TC1900011900.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900204978.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204978.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204978.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204978.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000198939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000544299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546361.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000546538.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000551747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000551747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000551747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000597261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000597261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000597261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000597261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000597261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000600432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000600432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000600432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | ENST00000600432.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011900.hg.1 | NM_006387.5 | Refseq |

CHERP
Genomic Map