Assay Details
| Gene Symbol: VWF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000552.4 | NP_000543.2 | 44 - 45 | 7807 | 140 | |
| GenBank mRNA | AK297600.1 | - | 16 - 17 | 2436 | 140 | |
| M10321.1 | - | 28 - 29 | 5336 | 140 | ||
| X04385.1 | - | 44 - 45 | 7651 | 140 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001130.hg.1
Set Membership: |
|

VWF
Genomic Map