Assay Details
| Gene Symbol: PSPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055490.1 | - | 1 | 2079 | 158 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | alkB homolog 7 | ALKBH7 |
ABH7, SPATA11, UNQ6002 |
Sense | Chr. 19 - 6372433 - 6375250 on NCBI Build GRCh38 | |
| Downstream | general transcription factor IIF subunit 1 | GTF2F1 |
BTF4, RAP74, TF2F1, TFIIF |
3.9kb | Anti-sense | Chr. 19 - 6379569 - 6393280 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PSPN
Genomic Map