Assay Details
| Gene Symbol: RFX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017027107.1 | XP_016882596.1 | 1 - 2 | 207 | 55 | |
| XM_017027108.1 | XP_016882597.1 | 1 - 2 | 207 | 55 | ||
| GenBank mRNA | AK303321.1 | - | 1 - 2 | 170 | 55 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK303321.1, XM_017027107.1, XM_017027108.1 transcripts.
Set Membership: |
|

RFX2
Genomic Map