Assay Details
Gene Symbol: DLGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277161.1 | NP_001264090.1 | 9 - 10 | 2650 | 99 | |
NM_004745.4 | NP_004736.2 | 10 - 11 | 2692 | 99 | ||
GenBank mRNA | AB000275.1 | - | 7 - 8 | 1247 | 99 | |
AF009204.1 | - | 8 - 9 | 2463 | 99 | ||
AF119817.1 | - | 10 - 11 | 2677 | 99 | ||
AF119818.1 | - | 10 - 11 | 2692 | 99 | ||
BC141862.1 | - | 9 - 10 | 2608 | 40027523 | 99 | |
KJ534823.1 | - | 9 - 10 | 2558 | 99 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000007.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08000118.hg.1 | TC08000007.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08000118.hg.1 | ENST00000356067.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08000118.hg.1 | ENST00000421627.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08000118.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144304.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144304.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144304.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144304.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144304.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144304.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144304.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144304.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800144304.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144304.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800144304.hg.1 | TC0800006475.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000421627.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000518530.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000520901.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000522499.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000524065.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | ENST00000612087.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_001277161.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0800006475.hg.1 | NM_004745.4 | Refseq |
