Assay Details
| Gene Symbol: MAP3K2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006609.4 | NP_006600.3 | 2 - 3 | 221 | 156 | |
| GenBank mRNA | AB208963.1 | - | 1 - 2 | 127 | 156 | |
| AF111105.1 | - | 2 - 3 | 222 | 156 | ||
| AF239798.1 | - | 1 - 2 | 152 | 156 | ||
| AK315007.1 | - | 3 - 4 | 405 | 156 | ||
| BC136293.1 | - | 2 - 3 | 190 | 9020280 | 156 | |
More Information
Updated Mapping Information
Set Membership: |
|

MAP3K2
Genomic Map