Assay Details
| Gene Symbol: USP25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013396.4 | NP_037528.3 | 18 - 19 | 2712 | 89 | |
| XM_017028324.1 | XP_016883813.1 | 18 - 19 | 2564 | 89 | ||
| XM_017028329.1 | XP_016883818.1 | 13 - 14 | 1724 | 89 | ||
| GenBank mRNA | AF134213.1 | - | 18 - 19 | 2542 | 89 | |
| AK022447.1 | - | 6 - 7 | 1014 | 89 | ||
| AK022574.1 | - | 9 - 10 | 1395 | 89 | ||
| BC075792.1 | - | 18 - 19 | 2713 | 30346400 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

USP25
Genomic Map