Assay Details
| Gene Symbol: MAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039538.1 | NP_001034627.1 | 2 - 3 | 273 | 105 | |
| GenBank mRNA | BC038857.1 | - | 2 - 3 | 262 | 105 | |
| BC066648.1 | - | 2 - 3 | 211 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC038857.1, BC066648.1, NM_001039538.1 transcripts.
Set Membership: |
|

MAP2
Genomic Map