Assay Details
| Gene Symbol: VPS29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282151.1 | NP_001269080.1 | 1 - 2 | 93 | 54 | |
| NM_016226.4 | NP_057310.1 | 1 - 2 | 93 | 54 | ||
| GenBank mRNA | AB618733.1 | - | 1 - 2 | 76 | 54 | |
| AF175264.1 | - | 1 - 2 | 62 | 54 | ||
| AF193795.1 | - | 1 - 2 | 55 | 54 | ||
| AK129767.1 | - | 1 - 2 | 55 | 54 | ||
| AK130510.1 | - | 891 | 54 | |||
| AL136614.1 | - | 1 - 2 | 71 | 54 | ||
| BC095446.1 | - | 1 - 2 | 64 | 30719438 | 54 | |
| BG707325.1 | - | 1 - 2 | 88 | 54 | ||
| CB158821.1 | - | 1 - 2 | 27 | 54 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB618733.1, AF175264.1, AF193795.1, AL136614.1, BC095446.1, NM_001282151.1, NM_016226.4 transcripts.
Set Membership: |
|

VPS29
Genomic Map