Assay Details
| Gene Symbol: MAGI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012850.1 | XP_016868339.1 | 23 - 24 | 4408 | 100 | |
| XM_017012851.1 | XP_016868340.1 | 23 - 24 | 4430 | 100 | ||
More Information
Set Membership: |
|

MAGI2
Genomic Map