Assay Details
| Gene Symbol: LAMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL133595.1 | - | 2 - 3 | 157 | 63 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000355452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000361069.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000462567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | ENST00000480883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008980.hg.1 | NM_006059.3 | Refseq |

LAMC3
Genomic Map