Assay Details
| Gene Symbol: ZCCHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318148.1 | NP_001305077.1 | 6 - 7 | 800 | 132 | |
| NM_024936.2 | NP_079212.2 | 6 - 7 | 800 | 132 | ||
| XM_011513835.1 | XP_011512137.1 | 6 - 7 | 767 | 132 | ||
| XM_017008128.1 | XP_016863617.1 | 5 - 6 | 679 | 132 | ||
| GenBank mRNA | AK299211.1 | - | 6 - 7 | 790 | 132 | |
| AK309675.1 | - | 6 - 7 | 789 | 132 | ||
| AY629351.1 | - | 6 - 7 | 793 | 132 | ||
| BC157834.1 | - | 6 - 7 | 794 | 9057053 | 132 | |
| BC171821.1 | - | 6 - 7 | 794 | 9054475 | 132 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

ZCCHC4
Genomic Map