Assay Details
| Gene Symbol: DNAJB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317099.1 | NP_001304028.1 | 1 - 2 | 138 | 131 | |
| GenBank mRNA | BI831822.1 | - | 1 - 2 | 137 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001317099.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000426517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000426517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000426517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000426517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000477671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000477671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000477671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000477671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000477671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000484662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000484662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000484662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000484662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000370763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000426517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000426517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000426517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000426517.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000476396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000477671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000477671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000477671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000477671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000477671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000484662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000484662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000484662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000484662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | ENST00000487931.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | NM_007034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018238.hg.1 | NM_007034.4 | Refseq |

DNAJB4
Genomic Map