Assay Details
| Gene Symbol: MED8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001653.2 | NP_001001653.1 | 4 - 5 | 498 | 86 | |
| NM_052877.4 | NP_443109.2 | 4 - 5 | 455 | 86 | ||
| NM_201542.4 | NP_963836.2 | 4 - 5 | 455 | 86 | ||
| GenBank mRNA | AF521562.1 | - | 3 - 4 | 407 | 86 | |
| BC010019.2 | - | 4 - 5 | 470 | 2905623 | 86 | |
| BC010543.1 | - | 3 - 4 | 396 | 86 | ||
| BG620658.1 | - | 4 - 5 | 456 | 86 | ||
| BI488866.1 | - | 3 - 4 | 411 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002584.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040201.hg.1 | TC01002584.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040201.hg.1 | ENST00000290663.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040201.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040201.hg.1 | ENST00000372457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040201.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040201.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040203.hg.1 | TC01002584.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040203.hg.1 | ENST00000290663.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040203.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040203.hg.1 | ENST00000372457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040203.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01040203.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204156.hg.1 | TC0100013925.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013925.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | ENST00000290663.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | ENST00000372455.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | ENST00000372457.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | ENST00000460803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | ENST00000473560.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | NM_001001653.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | NM_052877.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | NM_201542.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100204158.hg.1 | TC0100013925.hg.1 | Clariom D TC |

MED8
Genomic Map