| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014289.hg.1 |
ENST00000322282.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014289.hg.1 |
ENST00000450171.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014289.hg.1 |
ENST00000456860.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014289.hg.1 |
ENST00000529432.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014289.hg.1 |
ENST00000529750.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014289.hg.1 |
ENST00000539133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014289.hg.1 |
NM_020716.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11014289.hg.1 |
TC11001124.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000322282.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000450171.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000456860.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000525945.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000529432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000529750.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000532581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
NM_001286563.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
NM_001286564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
NM_020716.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000322282.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000450171.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000456860.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000525945.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000529432.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000529750.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
ENST00000532581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
NM_001286563.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
NM_001286564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
NM_020716.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100202867.hg.1 |
TC1100013109.hg.1 |
Clariom D TC |