Assay Details
| Gene Symbol: ENO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242699.1 | NP_001229628.1 | 7 - 8 | 1043 | 96 | |
| XM_005269815.3 | XP_005269872.1 | 7 - 8 | 2325 | 96 | ||
| XM_006717835.3 | XP_006717898.1 | 7 - 8 | 2325 | 96 | ||
| XM_006717836.3 | XP_006717899.1 | 6 - 7 | 860 | 96 | ||
| XM_011539797.2 | XP_011538099.1 | 7 - 8 | 2325 | 96 | ||
| XM_011539799.2 | XP_011538101.1 | 7 - 8 | 2325 | 96 | ||
| XM_017016241.1 | XP_016871730.1 | 7 - 8 | 2325 | 96 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000848.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10010297.hg.1 | ENST00000341276.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010297.hg.1 | ENST00000369207.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010297.hg.1 | ENST00000409522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010297.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10010297.hg.1 | TC10000848.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000512864.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009012.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161756.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161756.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161756.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161756.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161756.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161756.hg.1 | ENST00000341276.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161756.hg.1 | ENST00000369207.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161756.hg.1 | ENST00000409522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161756.hg.1 | ENST00000622726.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000161756.hg.1 | NM_001242699.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000161756.hg.1 | TC1000009012.hg.1 | Clariom D TC |

ENO4
Genomic Map