Assay Details
| Gene Symbol: HSPA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278205.1 | NP_001265134.1 | 3 - 4 | 470 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000378372.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000441647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000470430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | ENST00000493863.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NM_016299.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000006858.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146932.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000146932.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146932.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146932.hg.1 | ENST00000437161.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000146932.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146932.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146932.hg.1 | TC1000006858.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000146933.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146933.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146933.hg.1 | NM_001278205.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146933.hg.1 | NR_103464.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000146933.hg.1 | TC1000006858.hg.1 | Clariom D TC |

HSPA14
Genomic Map