Assay Details
| Gene Symbol: SLC9A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173653.3 | NP_775924.1 | 1 - 2 | 377 | 85 | |
| XM_011512704.2 | XP_011511006.1 | 1 - 2 | 350 | 85 | ||
| XM_017006202.1 | XP_016861691.1 | 1 - 2 | 350 | 85 | ||
| GenBank mRNA | AB089794.1 | - | 1 - 2 | 168 | 85 | |
| AK315107.1 | - | 1 - 2 | 304 | 85 | ||
| AL832304.1 | - | 1 - 2 | 242 | 85 | ||
| AY254100.1 | - | 1 - 2 | 292 | 85 | ||
| BC035779.1 | - | 1 - 2 | 293 | 5590356 | 85 | |
| DA387367.1 | - | 1 - 2 | 350 | 85 | ||
| DB285312.1 | - | 1 - 2 | 378 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001862.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03033624.hg.1 | TC03001862.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033624.hg.1 | ENST00000316549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033624.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033618.hg.1 | TC03001862.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033618.hg.1 | ENST00000316549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033618.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033620.hg.1 | TC03001862.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033620.hg.1 | ENST00000316549.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033620.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033620.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194840.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194840.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194840.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194840.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194840.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194840.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194840.hg.1 | TC0300012686.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300194847.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194847.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194847.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194847.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194847.hg.1 | TC0300012686.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300194843.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194843.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194843.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194843.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194843.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194843.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194843.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194843.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194843.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194843.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194843.hg.1 | TC0300012686.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300194842.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194842.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194842.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194842.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194842.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194842.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194842.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194842.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194842.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194842.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194842.hg.1 | TC0300012686.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 216190 | - | - |

SLC9A9
Genomic Map