Assay Details
| Gene Symbol: SLC9A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017006203.1 | XP_016861692.1 | 1 - 2 | 345 | 86 | |
| GenBank mRNA | AK301816.1 | - | 1 - 2 | 313 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK301816.1, XM_017006203.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000316549.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000474727.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000483124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | ENST00000498717.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012686.hg.1 | NM_173653.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 198279 | - | - |

SLC9A9
Genomic Map