Assay Details
| Gene Symbol: TSPAN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012338.3 | NP_036470.1 | 1 - 2 | 310 | 86 | |
| XM_005250239.2 | XP_005250296.1 | 2 - 3 | 311 | 86 | ||
| XM_017011912.1 | XP_016867401.1 | 2 - 3 | 216 | 86 | ||
| XM_017011913.1 | XP_016867402.1 | 1 - 2 | 333 | 86 | ||
| GenBank mRNA | AK312239.1 | - | 1 - 2 | 149 | 86 | |
| AY358703.1 | - | 1 - 2 | 143 | 86 | ||
| BC031265.1 | - | 1 - 2 | 308 | 5275953 | 86 | |
| BI964728.1 | - | 1 - 2 | 311 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001804.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312239.1,AY358703.1,BC031265.1,NM_012338.3,XM_005250239.2,XM_017011912.1,XM_017011913.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07027963.hg.1 | TC07001804.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027963.hg.1 | ENST00000222747.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027963.hg.1 | ENST00000415871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027963.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027966.hg.1 | TC07001804.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027966.hg.1 | ENST00000222747.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027966.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027958.hg.1 | TC07001804.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027958.hg.1 | ENST00000222747.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027958.hg.1 | ENST00000415871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027958.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189398.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189398.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189398.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189398.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189398.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189398.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189398.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189398.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189398.hg.1 | TC0700012416.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700189399.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189399.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189399.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189399.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189399.hg.1 | TC0700012416.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189393.hg.1 | TC0700012416.hg.1 | Clariom D TC |

TSPAN12
Genomic Map