Assay Details
| Gene Symbol: TSPAN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012338.3 | NP_036470.1 | 5 - 6 | 740 | 71 | |
| XM_005250239.2 | XP_005250296.1 | 6 - 7 | 741 | 71 | ||
| XM_011515993.1 | XP_011514295.1 | 5 - 6 | 557 | 71 | ||
| XM_011515994.1 | XP_011514296.1 | 5 - 6 | 475 | 71 | ||
| XM_017011912.1 | XP_016867401.1 | 6 - 7 | 646 | 71 | ||
| GenBank mRNA | AF124522.1 | - | 4 - 5 | 431 | 71 | |
| AK312239.1 | - | 5 - 6 | 579 | 71 | ||
| AY358703.1 | - | 5 - 6 | 573 | 71 | ||
| BC031265.1 | - | 5 - 6 | 738 | 5275953 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001804.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07027950.hg.1 | TC07001804.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027950.hg.1 | ENST00000222747.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027950.hg.1 | ENST00000415871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07027950.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189383.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189383.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189383.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189383.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189383.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189383.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189383.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189383.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189383.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700189383.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700189383.hg.1 | TC0700012416.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000222747.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000415871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000424710.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000430985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000433758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000441017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | ENST00000450414.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012416.hg.1 | NM_012338.3 | Refseq |

TSPAN12
Genomic Map